Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCF2L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MCF2L2 - MCF.2 cell line derived transforming sequence-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015078.3 | 3830 | UTR 3 | NP_055893.3 | |||
XM_011512585.2 | 3830 | UTR 3 | XP_011510887.1 | |||
XM_011512586.2 | 3830 | UTR 3 | XP_011510888.1 | |||
XM_017005943.1 | 3830 | Intron | XP_016861432.1 | |||
XM_017005944.1 | 3830 | Intron | XP_016861433.1 | |||
XM_017005945.1 | 3830 | Intron | XP_016861434.1 | |||
XM_017005946.1 | 3830 | Intron | XP_016861435.1 |
Set Membership: |
HapMap |