Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610326 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNASEH2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNASEH2B - ribonuclease H2 subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142279.2 | Intron | NP_001135751.1 | ||||
NM_024570.3 | Intron | NP_078846.2 | ||||
XM_006719867.3 | Intron | XP_006719930.1 | ||||
XM_011535229.1 | Intron | XP_011533531.1 | ||||
XM_011535230.2 | Intron | XP_011533532.1 | ||||
XM_011535231.2 | Intron | XP_011533533.1 | ||||
XM_011535233.2 | Intron | XP_011533535.1 | ||||
XM_017020747.1 | Intron | XP_016876236.1 |