Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATTGCTGTGCAATAGATCTTGG[A/G]AACTTATTCCTCCTGTCTAACCGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM151B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM151B - family with sequence similarity 151 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_205548.2 | Intron | NP_991111.2 | ||||
XM_006714564.3 | Intron | XP_006714627.2 | ||||
XM_011543234.2 | Intron | XP_011541536.1 | ||||
XM_011543235.2 | Intron | XP_011541537.1 | ||||
XM_011543236.2 | Intron | XP_011541538.1 | ||||
XM_011543237.2 | Intron | XP_011541539.1 | ||||
XM_017009166.1 | Intron | XP_016864655.1 | ||||
XM_017009167.1 | Intron | XP_016864656.1 | ||||
XM_017009168.1 | Intron | XP_016864657.1 | ||||
XM_017009169.1 | Intron | XP_016864658.1 | ||||
XM_017009170.1 | Intron | XP_016864659.1 |