Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614956 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLFN12L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLFN12L - schlafen family member 12 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195790.1 | 7899 | Missense Mutation | TAT,TGT | Y,C 383 | NP_001182719.1 | |
XM_017024005.1 | 7899 | Missense Mutation | TAT,TGT | Y,C 412 | XP_016879494.1 | |
XM_017024006.1 | 7899 | Missense Mutation | TAT,TGT | Y,C 383 | XP_016879495.1 | |
XM_017024007.1 | 7899 | Missense Mutation | TAT,TGT | Y,C 383 | XP_016879496.1 | |
XM_017024008.1 | 7899 | Missense Mutation | TAT,TGT | Y,C 365 | XP_016879497.1 | |
XM_017024009.1 | 7899 | Missense Mutation | TAT,TGT | Y,C 365 | XP_016879498.1 |