Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGAATCTGGTTTAGACCCTAAC[C/G]CTGCTCAGCACCTGCTCTGGTGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163730 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOS2 - nitric oxide synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000625.4 | Intron | NP_000616.3 | ||||
XM_011524859.2 | Intron | XP_011523161.1 | ||||
XM_011524860.2 | Intron | XP_011523162.1 | ||||
XM_011524861.1 | Intron | XP_011523163.1 | ||||
XM_011524862.1 | Intron | XP_011523164.1 |
Set Membership: |
HapMap |