Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
55 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613439 MIM: 607055 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNST PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNST - consortin, connexin sorting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139459.1 | 226 | UTR 5 | NP_001132931.1 | |||
NM_152609.2 | 226 | Intron | NP_689822.2 | |||
XM_005273081.3 | 226 | Intron | XP_005273138.2 | |||
XM_005273083.4 | 226 | Intron | XP_005273140.1 | |||
XM_011544110.2 | 226 | Intron | XP_011542412.1 | |||
XM_011544111.1 | 226 | UTR 5 | XP_011542413.1 | |||
XM_011544112.1 | 226 | Intron | XP_011542414.1 | |||
XM_011544113.1 | 226 | UTR 5 | XP_011542415.1 | |||
XM_011544114.2 | 226 | Intron | XP_011542416.1 | |||
XM_017000487.1 | 226 | Intron | XP_016855976.1 | |||
XM_017000488.1 | 226 | Intron | XP_016855977.1 | |||
XM_017000489.1 | 226 | UTR 5 | XP_016855978.1 |
TFB2M - transcription factor B2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022366.2 | 226 | Intron | NP_071761.1 | |||
XM_011544248.2 | 226 | Intron | XP_011542550.1 | |||
XM_017002055.1 | 226 | Intron | XP_016857544.1 |
Set Membership: |
HapMap JSNP |