Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTAAAAAGTAGCTACTAAATGT[A/G]AAGTGGTACAGTAGAAATACTTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615283 MIM: 616086 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EXOC8 - exocyst complex component 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPRTN - SprT-like N-terminal domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010984.3 | Intron | NP_001010984.1 | ||||
NM_001261462.2 | Intron | NP_001248391.1 | ||||
NM_032018.6 | Intron | NP_114407.3 | ||||
XM_006711818.2 | Intron | XP_006711881.1 | ||||
XM_011544289.2 | Intron | XP_011542591.1 |