Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTCATGTGAAAAACTATGGCA[G/T]TTGTATCTGAGCATTCTGTAGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602744 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf131 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf131 - chromosome 1 open reading frame 131 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNPAT - glyceronephosphate O-acyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316350.1 | Intron | NP_001303279.1 | ||||
NM_014236.3 | Intron | NP_055051.1 | ||||
XM_005273313.4 | Intron | XP_005273370.1 | ||||
XM_011544303.2 | Intron | XP_011542605.1 | ||||
XM_011544304.2 | Intron | XP_011542606.1 |
Set Membership: |
HapMap |