Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR7A5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR7A5 - olfactory receptor family 7 subfamily A member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017506.1 | 1001 | Silent Mutation | TTC,TTT | F,F 288 | NP_059976.1 | |
XM_011527912.2 | 1001 | Silent Mutation | TTC,TTT | F,F 288 | XP_011526214.1 | |
XM_011527913.2 | 1001 | Silent Mutation | TTC,TTT | F,F 288 | XP_011526215.1 | |
XM_017026583.1 | 1001 | Silent Mutation | TTC,TTT | F,F 332 | XP_016882072.1 |
OR7C1 - olfactory receptor family 7 subfamily C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198944.1 | 1001 | Intron | NP_945182.1 | |||
XM_005259853.3 | 1001 | Intron | XP_005259910.1 | |||
XM_011527915.2 | 1001 | Intron | XP_011526217.1 |