Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608650 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ULK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ULK2 - unc-51 like autophagy activating kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142610.1 | 4794 | Intron | NP_001136082.1 | |||
NM_014683.3 | 4794 | UTR 3 | NP_055498.3 | |||
XM_011524087.2 | 4794 | UTR 3 | XP_011522389.1 | |||
XM_017025424.1 | 4794 | UTR 3 | XP_016880913.1 | |||
XM_017025425.1 | 4794 | Intron | XP_016880914.1 | |||
XM_017025426.1 | 4794 | UTR 3 | XP_016880915.1 | |||
XM_017025427.1 | 4794 | UTR 3 | XP_016880916.1 | |||
XM_017025428.1 | 4794 | UTR 3 | XP_016880917.1 |
Set Membership: |
HapMap |