Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600280 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NUBP1 - nucleotide binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278506.1 | 190 | Intron | NP_001265435.1 | |||
NM_001323594.1 | 190 | Intron | NP_001310523.1 | |||
NM_001323595.1 | 190 | Intron | NP_001310524.1 | |||
NM_001323596.1 | 190 | Intron | NP_001310525.1 | |||
NM_001323597.1 | 190 | Intron | NP_001310526.1 | |||
NM_002484.3 | 190 | Intron | NP_002475.2 | |||
XM_017023252.1 | 190 | UTR 5 | XP_016878741.1 |
TEKT5 - tektin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144674.1 | 190 | Intron | NP_653275.1 | |||
XM_011522383.2 | 190 | Intron | XP_011520685.1 | |||
XM_011522385.2 | 190 | Intron | XP_011520687.1 | |||
XM_011522389.1 | 190 | Intron | XP_011520691.1 | |||
XM_011522390.2 | 190 | Intron | XP_011520692.1 | |||
XM_017022955.1 | 190 | Intron | XP_016878444.1 | |||
XM_017022956.1 | 190 | Intron | XP_016878445.1 | |||
XM_017022957.1 | 190 | Intron | XP_016878446.1 | |||
XM_017022958.1 | 190 | Intron | XP_016878447.1 |
Set Membership: |
HapMap |