Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615143 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf78 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf78 - chromosome 9 open reading frame 78 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP20 - ubiquitin specific peptidase 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008563.4 | Intron | NP_001008563.2 | ||||
NM_001110303.3 | Intron | NP_001103773.2 | ||||
NM_006676.7 | Intron | NP_006667.3 | ||||
XM_005251665.3 | Intron | XP_005251722.1 | ||||
XM_005251666.3 | Intron | XP_005251723.1 | ||||
XM_011518161.2 | Intron | XP_011516463.1 | ||||
XM_011518162.2 | Intron | XP_011516464.1 | ||||
XM_017014221.1 | Intron | XP_016869710.1 |