Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600857 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDHA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SDHA - succinate dehydrogenase complex flavoprotein subunit A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294332.1 | Intron | NP_001281261.1 | ||||
NM_004168.3 | Intron | NP_004159.2 | ||||
XM_005248331.3 | Intron | XP_005248388.1 | ||||
XM_011514072.1 | Intron | XP_011512374.1 | ||||
XM_011514073.1 | Intron | XP_011512375.1 | ||||
XM_017009685.1 | Intron | XP_016865174.1 |