Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAACATAGCATGCTTGGGAGGAG[C/T]GAAGGAAACATTGAAGCCAAGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146660 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL7 - interleukin 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZC2HC1A - zinc finger C2HC-type containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016010.2 | Intron | NP_057094.2 | ||||
XM_005251256.1 | Intron | XP_005251313.1 | ||||
XM_006716454.2 | Intron | XP_006716517.1 | ||||
XM_011517539.2 | Intron | XP_011515841.1 |
Set Membership: |
HapMap |