Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGTCTTTTGTCTTTTGCTGTTA[C/T]CATCTTTACAGTACATTTCATGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FARP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FARP2 - FERM, ARH/RhoGEF and pleckstrin domain protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282983.1 | Intron | NP_001269912.1 | ||||
NM_001282984.1 | Intron | NP_001269913.1 | ||||
NM_014808.3 | Intron | NP_055623.1 | ||||
XM_005247050.3 | Intron | XP_005247107.1 | ||||
XM_011512233.1 | Intron | XP_011510535.1 | ||||
XM_011512234.1 | Intron | XP_011510536.1 | ||||
XM_011512235.1 | Intron | XP_011510537.1 | ||||
XM_011512236.1 | Intron | XP_011510538.1 | ||||
XM_017005416.1 | Intron | XP_016860905.1 | ||||
XM_017005417.1 | Intron | XP_016860906.1 | ||||
XM_017005418.1 | Intron | XP_016860907.1 | ||||
XM_017005419.1 | Intron | XP_016860908.1 | ||||
XM_017005420.1 | Intron | XP_016860909.1 |