Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCGAAGCCTGCAGCCCCCACCC[A/G]GCCGCCCAGACCCAGAGCTGAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609235 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRSK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRSK1 - BR serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032430.1 | 2522 | Missense Mutation | AGC,GGC | S,G 749 | NP_115806.1 | |
XM_005259327.3 | 2522 | Missense Mutation | AGC,GGC | S,G 659 | XP_005259384.1 | |
XM_011527395.2 | 2522 | Missense Mutation | AGC,GGC | S,G 573 | XP_011525697.2 |
TMEM150B - transmembrane protein 150B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085488.2 | 2522 | Intron | NP_001078957.1 | |||
NM_001282011.1 | 2522 | Intron | NP_001268940.1 | |||
XM_005258812.3 | 2522 | Intron | XP_005258869.1 | |||
XM_011526850.2 | 2522 | Intron | XP_011525152.1 | |||
XM_011526851.2 | 2522 | Intron | XP_011525153.1 | |||
XM_011526852.2 | 2522 | Intron | XP_011525154.1 | |||
XM_011526853.2 | 2522 | Intron | XP_011525155.1 | |||
XM_011526854.2 | 2522 | Intron | XP_011525156.1 | |||
XM_011526855.2 | 2522 | Intron | XP_011525157.1 | |||
XM_011526856.2 | 2522 | Intron | XP_011525158.1 | |||
XM_011526857.2 | 2522 | Intron | XP_011525159.1 | |||
XM_017026667.1 | 2522 | Intron | XP_016882156.1 | |||
XM_017026668.1 | 2522 | Intron | XP_016882157.1 |