Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTAGGGTAGGGTGAACAAATCTT[C/T]TGTCTCAGGAATTGCACACCCAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607723 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SUN1 - Sad1 and UNC84 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130965.2 | Intron | NP_001124437.1 | ||||
NM_001171944.1 | Intron | NP_001165415.1 | ||||
NM_001171945.1 | Intron | NP_001165416.1 | ||||
NM_001171946.1 | Intron | NP_001165417.1 | ||||
NM_025154.5 | Intron | NP_079430.3 |
Set Membership: |
HapMap |