Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616555 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL17B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARL17B - ADP ribosylation factor like GTPase 17B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC37A - leucine rich repeat containing 37A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014834.4 | Intron | NP_055649.4 | ||||
XM_005257879.4 | Intron | XP_005257936.1 | ||||
XM_005257880.4 | Intron | XP_005257937.1 | ||||
XM_005257882.4 | Intron | XP_005257939.1 | ||||
XM_005257883.4 | Intron | XP_005257940.1 | ||||
XM_005257884.4 | Intron | XP_005257941.1 | ||||
XM_006722210.3 | Intron | XP_006722273.1 | ||||
XM_006722211.3 | Intron | XP_006722274.1 | ||||
XM_011525539.2 | Intron | XP_011523841.1 | ||||
XM_017025472.1 | Intron | XP_016880961.1 | ||||
XM_017025473.1 | Intron | XP_016880962.1 |
Set Membership: |
HapMap |