Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM16L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRIM16L - tripartite motif containing 16-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037330.1 | Intron | NP_001032407.1 | ||||
XM_005256479.4 | Intron | XP_005256536.1 | ||||
XM_005256480.4 | Intron | XP_005256537.1 | ||||
XM_005256481.4 | Intron | XP_005256538.1 | ||||
XM_005256483.4 | Intron | XP_005256540.1 | ||||
XM_011523689.2 | Intron | XP_011521991.1 | ||||
XM_011523690.2 | Intron | XP_011521992.1 | ||||
XM_017024245.1 | Intron | XP_016879734.1 | ||||
XM_017024246.1 | Intron | XP_016879735.1 | ||||
XM_017024247.1 | Intron | XP_016879736.1 | ||||
XM_017024248.1 | Intron | XP_016879737.1 | ||||
XM_017024249.1 | Intron | XP_016879738.1 | ||||
XM_017024250.1 | Intron | XP_016879739.1 | ||||
XM_017024251.1 | Intron | XP_016879740.1 | ||||
XM_017024252.1 | Intron | XP_016879741.1 |