Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAGGTTGGGACTCTGGATGCG[C/T]GCGGGGCTCTGGCCTACCGGTGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609176 MIM: 610824 | ||||||||||||||||||||
Literature Links: |
PMF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PMF1 - polyamine-modulated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199653.1 | Intron | NP_001186582.1 | ||||
NM_001199654.1 | Intron | NP_001186583.1 | ||||
NM_007221.3 | Intron | NP_009152.2 |
PMF1-BGLAP - PMF1-BGLAP readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199661.1 | Intron | NP_001186590.1 | ||||
NM_001199662.1 | Intron | NP_001186591.1 | ||||
NM_001199663.1 | Intron | NP_001186592.1 | ||||
NM_001199664.1 | Intron | NP_001186593.1 |
SLC25A44 - solute carrier family 25 member 44 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |