Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191030 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP210L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUP210L - nucleoporin 210 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPM3 - tropomyosin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001043351.1 | 1362 | UTR 3 | NP_001036816.1 | |||
NM_001043352.1 | 1362 | UTR 3 | NP_001036817.1 | |||
NM_001043353.1 | 1362 | UTR 3 | NP_001036818.1 | |||
NM_001278188.1 | 1362 | UTR 3 | NP_001265117.1 | |||
NM_001278189.1 | 1362 | UTR 3 | NP_001265118.1 | |||
NM_001278190.1 | 1362 | UTR 3 | NP_001265119.1 | |||
NM_001278191.1 | 1362 | UTR 3 | NP_001265120.1 | |||
NM_152263.3 | 1362 | Intron | NP_689476.2 | |||
NM_153649.3 | 1362 | UTR 3 | NP_705935.1 |
Set Membership: |
HapMap |