Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCAGACAAAAGCAACCTTAATGA[G/A]TCTTCCTAACAAGGCTGGGGGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANTXRL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANTXRL - anthrax toxin receptor-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278688.2 | Intron | NP_001265617.1 | ||||
XM_006717684.3 | Intron | XP_006717747.1 | ||||
XM_011539430.2 | Intron | XP_011537732.1 | ||||
XM_011539431.2 | Intron | XP_011537733.1 | ||||
XM_011539433.2 | Intron | XP_011537735.1 | ||||
XM_011539434.2 | Intron | XP_011537736.1 | ||||
XM_011539437.2 | Intron | XP_011537739.1 | ||||
XM_011539438.2 | Intron | XP_011537740.1 | ||||
XM_011539439.2 | Intron | XP_011537741.1 | ||||
XM_011539440.2 | Intron | XP_011537742.1 | ||||
XM_011539441.1 | Intron | XP_011537743.1 | ||||
XM_011539442.1 | Intron | XP_011537744.1 | ||||
XM_011539443.1 | Intron | XP_011537745.1 | ||||
XM_017015818.1 | Intron | XP_016871307.1 | ||||
XM_017015819.1 | Intron | XP_016871308.1 |
LOC105378577 - basic proline-rich protein-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |