Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613593 MIM: 613594 | ||||||||||||||||||||
Literature Links: |
BTN3A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTN3A1 - butyrophilin subfamily 3 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145008.1 | 820 | Missense Mutation | GAA,GCA | E,A 151 | NP_001138480.1 | |
NM_001145009.1 | 820 | Missense Mutation | GAA,GCA | E,A 203 | NP_001138481.1 | |
NM_007048.5 | 820 | Missense Mutation | GAA,GCA | E,A 203 | NP_008979.3 | |
NM_194441.2 | 820 | Missense Mutation | GAA,GCA | E,A 203 | NP_919423.1 | |
XM_005248833.2 | 820 | Missense Mutation | GAA,GCA | E,A 203 | XP_005248890.1 | |
XM_005248834.3 | 820 | Missense Mutation | GAA,GCA | E,A 203 | XP_005248891.1 | |
XM_006714983.2 | 820 | Intron | XP_006715046.1 |