Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTCTTGTATGCCTGAACTAATT[A/C]CTCAGAGGCGAGACTGCTTTTCTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC17 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TTC17 - tetratricopeptide repeat domain 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307943.1 | Intron | NP_001294872.1 | ||||
NM_018259.5 | Intron | NP_060729.2 | ||||
XM_005253016.1 | Intron | XP_005253073.1 | ||||
XM_006718263.1 | Intron | XP_006718326.1 | ||||
XM_011520218.2 | Intron | XP_011518520.1 | ||||
XM_017018010.1 | Intron | XP_016873499.1 |