Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTGGTCCCCTGGAATGGGTGG[A/G]GTATGTGTGCCCTGGTTCCCTCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
35 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606351 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESPN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ESPN - espin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031475.2 | Intron | NP_113663.2 | ||||
XM_005263501.2 | Intron | XP_005263558.1 | ||||
XM_011542231.1 | Intron | XP_011540533.1 | ||||
XM_011542232.1 | Intron | XP_011540534.1 | ||||
XM_011542233.2 | Intron | XP_011540535.1 | ||||
XM_011542236.2 | Intron | XP_011540538.1 | ||||
XM_011542237.1 | Intron | XP_011540539.1 | ||||
XM_011542238.2 | Intron | XP_011540540.1 | ||||
XM_017002433.1 | Intron | XP_016857922.1 | ||||
XM_017002434.1 | Intron | XP_016857923.1 |
Set Membership: |
HapMap |