Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 103390 | ||||||||||||||||||||
Literature Links: |
AHNAK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AHNAK - AHNAK nucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001620.2 | Intron | NP_001611.1 | ||||
NM_024060.3 | Intron | NP_076965.2 | ||||
XM_005274240.1 | Intron | XP_005274297.1 | ||||
XM_005274241.2 | Intron | XP_005274298.1 | ||||
XM_005274245.1 | Intron | XP_005274302.1 | ||||
XM_017018270.1 | Intron | XP_016873759.1 | ||||
XM_017018271.1 | Intron | XP_016873760.1 | ||||
XM_017018272.1 | Intron | XP_016873761.1 | ||||
XM_017018273.1 | Intron | XP_016873762.1 |