Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTATGTCCACATAAATAAGTAAA[A/G]TATGATTTTTAGTAGCTGGTAATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613524 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4677 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4677 - microRNA 4677 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SDCCAG8 - serologically defined colon cancer antigen 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006642.3 | Intron | NP_006633.1 | ||||
XM_005273013.4 | Intron | XP_005273070.1 | ||||
XM_005273018.1 | Intron | XP_005273075.1 | ||||
XM_005273021.4 | Intron | XP_005273078.1 | ||||
XM_005273022.3 | Intron | XP_005273079.1 | ||||
XM_005273023.4 | Intron | XP_005273080.1 | ||||
XM_006711727.3 | Intron | XP_006711790.1 | ||||
XM_006711728.3 | Intron | XP_006711791.1 | ||||
XM_011544021.2 | Intron | XP_011542323.1 | ||||
XM_011544022.2 | Intron | XP_011542324.1 | ||||
XM_011544023.2 | Intron | XP_011542325.1 | ||||
XM_011544024.2 | Intron | XP_011542326.1 | ||||
XM_011544026.2 | Intron | XP_011542328.1 | ||||
XM_011544028.2 | Intron | XP_011542330.1 | ||||
XM_011544029.2 | Intron | XP_011542331.1 | ||||
XM_011544030.2 | Intron | XP_011542332.1 | ||||
XM_017000103.1 | Intron | XP_016855592.1 | ||||
XM_017000104.1 | Intron | XP_016855593.1 | ||||
XM_017000105.1 | Intron | XP_016855594.1 | ||||
XM_017000106.1 | Intron | XP_016855595.1 | ||||
XM_017000107.1 | Intron | XP_016855596.1 | ||||
XM_017000108.1 | Intron | XP_016855597.1 | ||||
XM_017000109.1 | Intron | XP_016855598.1 |