Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCACAGACACCCCTTATCAGGA[C/G]TACAAAGCCCGAGCTGTGTATTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615094 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PROX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PROX2 - prospero homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080408.2 | 2492 | UTR 3 | NP_001073877.2 | |||
NM_001243007.1 | 2492 | UTR 3 | NP_001229936.1 | |||
XM_005267543.3 | 2492 | Intron | XP_005267600.1 | |||
XM_017021222.1 | 2492 | Intron | XP_016876711.1 |
Set Membership: |
HapMap |