Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616226 MIM: 616605 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATG2B - autophagy related 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GSKIP - GSK3B interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271904.1 | Intron | NP_001258833.1 | ||||
NM_001271905.1 | Intron | NP_001258834.1 | ||||
NM_001271906.1 | Intron | NP_001258835.1 | ||||
NM_016472.4 | Intron | NP_057556.2 | ||||
XM_017021365.1 | Intron | XP_016876854.1 |