Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C14orf180 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C14orf180 - chromosome 14 open reading frame 180 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM179 - transmembrane protein 179 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286389.1 | 3242 | UTR 3 | NP_001273318.1 | |||
NM_001286390.1 | 3242 | Intron | NP_001273319.1 | |||
XM_011536745.2 | 3242 | UTR 3 | XP_011535047.1 | |||
XM_011536746.2 | 3242 | Intron | XP_011535048.1 | |||
XM_011536750.2 | 3242 | UTR 3 | XP_011535052.1 |