Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTGCGCACGTGTGAGTGGGAGC[A/G]TATGTGCACATGTGTGCATGTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614176 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFYVE28 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZFYVE28 - zinc finger FYVE-type containing 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172656.1 | Intron | NP_001166127.1 | ||||
NM_001172657.1 | Intron | NP_001166128.1 | ||||
NM_001172658.1 | Intron | NP_001166129.1 | ||||
NM_001172659.1 | Intron | NP_001166130.1 | ||||
NM_001172660.1 | Intron | NP_001166131.1 | ||||
NM_020972.2 | Intron | NP_066023.2 | ||||
XM_006713900.3 | Intron | XP_006713963.1 | ||||
XM_006713902.3 | Intron | XP_006713965.1 | ||||
XM_006713904.3 | Intron | XP_006713967.1 | ||||
XM_011513540.2 | Intron | XP_011511842.1 | ||||
XM_011513541.2 | Intron | XP_011511843.1 | ||||
XM_017008502.1 | Intron | XP_016863991.1 | ||||
XM_017008503.1 | Intron | XP_016863992.1 | ||||
XM_017008504.1 | Intron | XP_016863993.1 | ||||
XM_017008505.1 | Intron | XP_016863994.1 | ||||
XM_017008506.1 | Intron | XP_016863995.1 | ||||
XM_017008507.1 | Intron | XP_016863996.1 | ||||
XM_017008508.1 | Intron | XP_016863997.1 | ||||
XM_017008509.1 | Intron | XP_016863998.1 | ||||
XM_017008510.1 | Intron | XP_016863999.1 |
Set Membership: |
HapMap |