Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191311 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDR2 - discoidin domain receptor tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014796.1 | Intron | NP_001014796.1 | ||||
NM_006182.2 | Intron | NP_006173.2 | ||||
XM_006711344.3 | Intron | XP_006711407.1 | ||||
XM_011509586.2 | Intron | XP_011507888.1 | ||||
XM_011509587.2 | Intron | XP_011507889.1 | ||||
XM_011509588.2 | Intron | XP_011507890.1 |
Set Membership: |
HapMap |