Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCATGACAAACACCTCCCCCCAA[A/C]CCCCACCCACCTTGTGAGTGCTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609238 | ||||||||||||||||||||
Literature Links: |
LOC102724601 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC102724601 - uncharacterized LOC102724601 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RABGAP1L - RAB GTPase activating protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035230.2 | Intron | NP_001030307.1 | ||||
NM_001243763.1 | Intron | NP_001230692.1 | ||||
NM_001243764.1 | Intron | NP_001230693.1 | ||||
NM_001243765.1 | Intron | NP_001230694.1 | ||||
NM_014857.4 | Intron | NP_055672.3 | ||||
XM_005245680.1 | Intron | XP_005245737.1 | ||||
XM_005245681.2 | Intron | XP_005245738.1 | ||||
XM_006711693.1 | Intron | XP_006711756.1 | ||||
XM_011510223.2 | Intron | XP_011508525.1 | ||||
XM_017002994.1 | Intron | XP_016858483.1 | ||||
XM_017002995.1 | Intron | XP_016858484.1 | ||||
XM_017002996.1 | Intron | XP_016858485.1 | ||||
XM_017002997.1 | Intron | XP_016858486.1 | ||||
XM_017002998.1 | Intron | XP_016858487.1 | ||||
XM_017002999.1 | Intron | XP_016858488.1 |