Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACCATCATCAAAATACCCCTTT[C/T]TTTGTTAGAAAAGTATTTTGTTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102576 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACVR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACVR1 - activin A receptor type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105.4 | Intron | NP_001096.1 | ||||
NM_001111067.2 | Intron | NP_001104537.1 | ||||
XM_005246939.3 | Intron | XP_005246996.1 | ||||
XM_005246940.3 | Intron | XP_005246997.1 | ||||
XM_006712825.3 | Intron | XP_006712888.1 | ||||
XM_011512106.2 | Intron | XP_011510408.1 | ||||
XM_011512107.2 | Intron | XP_011510409.1 | ||||
XM_011512108.2 | Intron | XP_011510410.1 |