Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615664 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TESPA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TESPA1 - thymocyte expressed, positive selection associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098815.2 | Intron | NP_001092285.1 | ||||
NM_001136030.2 | Intron | NP_001129502.1 | ||||
NM_001261844.1 | Intron | NP_001248773.1 | ||||
NM_014796.2 | Intron | NP_055611.1 | ||||
XM_005269247.2 | Intron | XP_005269304.1 | ||||
XM_006719715.3 | Intron | XP_006719778.1 | ||||
XM_011539035.2 | Intron | XP_011537337.1 | ||||
XM_011539036.1 | Intron | XP_011537338.1 | ||||
XM_011539037.2 | Intron | XP_011537339.1 | ||||
XM_017020262.1 | Intron | XP_016875751.1 | ||||
XM_017020263.1 | Intron | XP_016875752.1 |