Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604543 MIM: 606569 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIMD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIMD1 - LIM domains containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIMD1-AS1 - LIMD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SACM1L - SAC1 suppressor of actin mutations 1-like (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319072.1 | Intron | NP_001306001.1 | ||||
NM_001319073.1 | Intron | NP_001306002.1 | ||||
NM_014016.4 | Intron | NP_054735.3 | ||||
XM_011533500.2 | Intron | XP_011531802.1 |