Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTTAACAGGATATTTTTACTT[A/T]TCTGAAAATTTTATAGCATACTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BANK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BANK1 - B-cell scaffold protein with ankyrin repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083907.2 | Intron | NP_001077376.2 | ||||
NM_001127507.2 | Intron | NP_001120979.2 | ||||
NM_017935.4 | Intron | NP_060405.4 | ||||
XM_017008337.1 | Intron | XP_016863826.1 |