Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGCGCCTTGCGGCCCAGGCCCG[C/G]AACCTTCCCTGGTCGTGCGCCCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603377 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC553103 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC553103 - uncharacterized LOC553103 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3936 - microRNA 3936 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A5 - solute carrier family 22 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308122.1 | 58 | UTR 5 | NP_001295051.1 | |||
NM_003060.3 | 58 | UTR 5 | NP_003051.1 | |||
XM_011543590.2 | 58 | Intron | XP_011541892.1 | |||
XM_017009778.1 | 58 | Intron | XP_016865267.1 |
Set Membership: |
DME Validated Inventoried |