Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123810 MIM: 615257 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CREB1 - cAMP responsive element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320793.1 | Intron | NP_001307722.1 | ||||
NM_004379.4 | Intron | NP_004370.1 | ||||
NM_134442.4 | Intron | NP_604391.1 | ||||
XM_011510645.1 | Intron | XP_011508947.1 | ||||
XM_011510646.2 | Intron | XP_011508948.1 | ||||
XM_011510647.2 | Intron | XP_011508949.1 | ||||
XM_011510648.2 | Intron | XP_011508950.1 | ||||
XM_011510649.2 | Intron | XP_011508951.1 | ||||
XM_011510650.2 | Intron | XP_011508952.1 | ||||
XM_011510651.2 | Intron | XP_011508953.1 | ||||
XM_017003399.1 | Intron | XP_016858888.1 | ||||
XM_017003400.1 | Intron | XP_016858889.1 | ||||
XM_017003401.1 | Intron | XP_016858890.1 |
METTL21A - methyltransferase like 21A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005246336.3 | Intron | XP_005246393.1 | ||||
XM_005246337.4 | Intron | XP_005246394.1 | ||||
XM_005246338.4 | Intron | XP_005246395.1 | ||||
XM_005246339.3 | Intron | XP_005246396.1 | ||||
XM_005246340.3 | Intron | XP_005246397.1 | ||||
XM_005246341.2 | Intron | XP_005246398.1 | ||||
XM_006712327.3 | Intron | XP_006712390.1 | ||||
XM_006712329.3 | Intron | XP_006712392.1 | ||||
XM_011510724.2 | Intron | XP_011509026.1 | ||||
XM_011510725.2 | Intron | XP_011509027.1 | ||||
XM_011510727.2 | Intron | XP_011509029.1 | ||||
XM_011510728.2 | Intron | XP_011509030.1 | ||||
XM_011510729.2 | Intron | XP_011509031.1 | ||||
XM_017003442.1 | Intron | XP_016858931.1 | ||||
XM_017003443.1 | Intron | XP_016858932.1 | ||||
XM_017003444.1 | Intron | XP_016858933.1 | ||||
XM_017003445.1 | Intron | XP_016858934.1 | ||||
XM_017003446.1 | Intron | XP_016858935.1 | ||||
XM_017003447.1 | Intron | XP_016858936.1 | ||||
XM_017003448.1 | Intron | XP_016858937.1 | ||||
XM_017003449.1 | Intron | XP_016858938.1 | ||||
XM_017003450.1 | Intron | XP_016858939.1 |