Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTCTGTGAGGCCTGCCCAGTTTC[A/T]CAGGTGAGAGTTACCCACTTTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM45B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM45B - transmembrane protein 45B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138788.3 | 84 | Intron | NP_620143.1 | |||
XM_005271400.4 | 84 | Intron | XP_005271457.1 | |||
XM_011542586.2 | 84 | Intron | XP_011540888.1 | |||
XM_011542588.2 | 84 | Intron | XP_011540890.1 | |||
XM_017017188.1 | 84 | Intron | XP_016872677.1 | |||
XM_017017189.1 | 84 | UTR 5 | XP_016872678.1 |