Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136130 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FMO1 - flavin containing monooxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282692.1 | Intron | NP_001269621.1 | ||||
NM_001282693.1 | Intron | NP_001269622.1 | ||||
NM_001282694.1 | Intron | NP_001269623.1 | ||||
NM_002021.2 | Intron | NP_002012.1 | ||||
XM_005245037.3 | Intron | XP_005245094.1 | ||||
XM_005245038.3 | Intron | XP_005245095.1 | ||||
XM_006711241.3 | Intron | XP_006711304.1 | ||||
XM_006711242.3 | Intron | XP_006711305.1 |
Set Membership: |
HapMap |