Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604911 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CNOT4 - CCR4-NOT transcription complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008225.2 | 3558 | Intron | NP_001008226.1 | |||
NM_001190847.1 | 3558 | UTR 3 | NP_001177776.1 | |||
NM_001190848.1 | 3558 | Intron | NP_001177777.1 | |||
NM_001190849.1 | 3558 | UTR 3 | NP_001177778.1 | |||
NM_001190850.1 | 3558 | UTR 3 | NP_001177779.1 | |||
NM_013316.3 | 3558 | UTR 3 | NP_037448.2 | |||
XM_017012235.1 | 3558 | UTR 3 | XP_016867724.1 |
Set Membership: |
HapMap |