Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACGGTTTAGCTGCCTCTGCTCA[A/G]CTCTTCCTCAAGAAGGGGACACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604309 | ||||||||||||||||||||
Literature Links: |
C7orf73 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C7orf73 - chromosome 7 open reading frame 73 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC13A4 - solute carrier family 13 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318192.1 | Intron | NP_001305121.1 | ||||
NM_012450.3 | Intron | NP_036582.2 | ||||
XM_011516024.2 | Intron | XP_011514326.1 | ||||
XM_017011962.1 | Intron | XP_016867451.1 | ||||
XM_017011963.1 | Intron | XP_016867452.1 |