Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGAGAATTCACAGAAACATTCA[C/T]GAGATTAGGCTTGCATTTCCTGTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604309 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf73 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C7orf73 - chromosome 7 open reading frame 73 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC13A4 - solute carrier family 13 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318192.1 | Intron | NP_001305121.1 | ||||
NM_012450.3 | Intron | NP_036582.2 | ||||
XM_011516024.2 | Intron | XP_011514326.1 | ||||
XM_017011962.1 | Intron | XP_016867451.1 | ||||
XM_017011963.1 | Intron | XP_016867452.1 |
Set Membership: |
HapMap |