Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604707 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR1B10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKR1B10 - aldo-keto reductase family 1 member B10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020299.4 | 395 | Silent Mutation | CTC,CTT | L,L 25 | NP_064695.3 | |
XM_011516416.1 | 395 | Silent Mutation | CTC,CTT | L,L 25 | XP_011514718.1 | |
XM_011516417.2 | 395 | Silent Mutation | CTC,CTT | L,L 25 | XP_011514719.1 |
Set Membership: |
HapMap |