Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCACCGCCACCTCGTGGATCTCA[A/G]AGCTGCAGGACGGCTCCAAACACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610855 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD26 - ankyrin repeat domain 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256053.1 | 19133 | Intron | NP_001242982.1 | |||
NM_014915.2 | 19133 | Intron | NP_055730.2 | |||
XM_006717423.2 | 19133 | Intron | XP_006717486.1 | |||
XM_006717425.3 | 19133 | Intron | XP_006717488.1 | |||
XM_011519416.2 | 19133 | Intron | XP_011517718.1 | |||
XM_017015928.1 | 19133 | Intron | XP_016871417.1 | |||
XM_017015929.1 | 19133 | UTR 3 | XP_016871418.1 | |||
XM_017015930.1 | 19133 | Intron | XP_016871419.1 | |||
XM_017015931.1 | 19133 | Intron | XP_016871420.1 | |||
XM_017015932.1 | 19133 | Intron | XP_016871421.1 | |||
XM_017015933.1 | 19133 | Intron | XP_016871422.1 | |||
XM_017015934.1 | 19133 | Intron | XP_016871423.1 |
LINC00202-1 - long intergenic non-protein coding RNA 202-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |