Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609642 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TARP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TARP - TCR gamma alternate reading frame protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003799.1 | Intron | NP_001003799.1 | ||||
NM_001003806.1 | Intron | NP_001003806.1 | ||||
XM_011515407.2 | Intron | XP_011513709.1 | ||||
XM_017012226.1 | Intron | XP_016867715.1 | ||||
XM_017012227.1 | Intron | XP_016867716.1 | ||||
XM_017012228.1 | Intron | XP_016867717.1 | ||||
XM_017012229.1 | Intron | XP_016867718.1 | ||||
XM_017012230.1 | Intron | XP_016867719.1 |