Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300427 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLGN4X PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NLGN4X - neuroligin 4, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282145.1 | 5132 | Intron | NP_001269074.1 | |||
NM_001282146.1 | 5132 | UTR 3 | NP_001269075.1 | |||
NM_020742.3 | 5132 | Intron | NP_065793.1 | |||
NM_181332.2 | 5132 | Intron | NP_851849.1 | |||
XM_005274564.2 | 5132 | Intron | XP_005274621.1 | |||
XM_005274565.2 | 5132 | UTR 3 | XP_005274622.1 | |||
XM_005274566.4 | 5132 | Intron | XP_005274623.1 | |||
XM_006724504.3 | 5132 | UTR 3 | XP_006724567.1 | |||
XM_011545547.2 | 5132 | UTR 3 | XP_011543849.1 | |||
XM_011545548.2 | 5132 | Intron | XP_011543850.1 | |||
XM_017029690.1 | 5132 | UTR 3 | XP_016885179.1 | |||
XM_017029691.1 | 5132 | UTR 3 | XP_016885180.1 | |||
XM_017029692.1 | 5132 | Intron | XP_016885181.1 | |||
XM_017029693.1 | 5132 | UTR 3 | XP_016885182.1 |
Set Membership: |
HapMap |