Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGAATTGCCTGGGACCCGTGA[A/G]CCCTTTTTTCCTTCCCCTTTCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614148 MIM: 602241 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF9B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1QTNF9B - C1q and tumor necrosis factor related protein 9B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1QTNF9B-AS1 - C1QTNF9B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014442.2 | Intron | NP_001014442.2 | ||||
NM_001135816.1 | Intron | NP_001129288.1 |
MIPEP - mitochondrial intermediate peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005932.3 | Intron | NP_005923.2 | ||||
XM_011535097.2 | Intron | XP_011533399.1 | ||||
XM_011535098.2 | Intron | XP_011533400.1 |
Set Membership: |
HapMap |