Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607519 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PARP4 - poly(ADP-ribose) polymerase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006437.3 | 4972 | Missense Mutation | CCG,CTG | P,L 1550 | NP_006428.2 | |
XM_011534931.1 | 4972 | Missense Mutation | CCG,CTG | P,L 1597 | XP_011533233.1 | |
XM_011534932.2 | 4972 | Missense Mutation | CCG,CTG | P,L 1550 | XP_011533234.1 |
Set Membership: |
HapMap |